missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorobutane CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
CHROMASOLV™, for HPLC,≥99.8%
110.00 € - 412.00 €
Chemical Identifiers
CAS | 109-69-3 |
---|---|
Molecular Formula | C4H9Cl |
Molecular Weight (g/mol) | 92.57 |
MDL Number | MFCD00001009 |
InChI Key | VFWCMGCRMGJXDK-UHFFFAOYSA-N |
Synonym | butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane |
PubChem CID | 8005 |
IUPAC Name | 1-chlorobutane |
SMILES | CCCCCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15694720
|
Honeywell-Chromasolv
34958-1L |
1 L | |||||||
15614730
|
Honeywell-Chromasolv
34958-2L |
2 L |
265.19 €
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15604730
|
Honeywell-Chromasolv
34958-2.5L |
2.5 L | |||||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
109-69-3 | |
92.57 | |
VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
8005 | |
CCCCCl |
C4H9Cl | |
MFCD00001009 | |
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
1-chlorobutane |
Specifications
109-69-3 | |
0.886g/mL at 25°C (lit.) | |
C4H9Cl | |
n20/D 1.402 | |
1730909 | |
VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
1-chlorobutane | |
8005 | |
80.1mmHg (78.4°C) | |
CHROMASOLV™ |
-123°C | |
77°C to 78°C | |
CH3(CH2)3Cl | |
MFCD00001009 | |
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
CCCCCl | |
92.57 | |
92.57g/mol | |
≥99.8% | |
1-Chlorobutane, for HPLC |
Safety and Handling
P210-P273-P301 + P310-P331-P403 + P235
H225-H304-H336-H412
missing translation for 'einecsNumber' : 203-696-6
missing translation for 'rtecsNumber' : EJ6300000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Chlorobutane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.