Learn More
1-Butylboronic acid, 98%, Thermo Scientific Chemicals
27.84 € - 210.05 €
Chemical Identifiers
CAS | 4426-47-5 |
---|---|
Molecular Formula | C4H11BO2 |
Molecular Weight (g/mol) | 101.94 |
MDL Number | MFCD00002106 |
InChI Key | QPKFVRWIISEVCW-UHFFFAOYSA-N |
Synonym | n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane |
PubChem CID | 20479 |
IUPAC Name | butylboronic acid |
SMILES | CCCCB(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484734
|
Thermo Scientific Alfa Aesar
A13725.03 |
1 g |
27.84 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494734
|
Thermo Scientific Alfa Aesar
A13725.06 |
5 g |
63.72 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404744
|
Thermo Scientific Alfa Aesar
A13725.14 |
25 g |
210.05 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Butylboronic acid is used as a reagent for preparation of volatile derivatives of diols, e.g. carbohydrates, for GC and mass spectrometry and to prepare chiral oxazaborolidines. It is a precursor to unsymmetric borinic acids, which are inhibitors of serine proteases. Can be used as a transport carrier in membrane-based sugar separations and as an analytical reagent in the determination of serum glucose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Butylboronic acid is used as a reagent for preparation of volatile derivatives of diols, e.g. carbohydrates, for GC and mass spectrometry and to prepare chiral oxazaborolidines. It is a precursor to unsymmetric borinic acids, which are inhibitors of serine proteases. Can be used as a transport carrier in membrane-based sugar separations and as an analytical reagent in the determination of serum glucose.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is hygroscopic in nature. Incompatible with oxidizing agents. Store under dry inert gas.
Chemical Identifiers
4426-47-5 | |
101.94 | |
QPKFVRWIISEVCW-UHFFFAOYSA-N | |
20479 | |
CCCCB(O)O |
C4H11BO2 | |
MFCD00002106 | |
n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane | |
butylboronic acid |
Specifications
4426-47-5 | |
C4H11BO2 | |
1733489 | |
n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane | |
QPKFVRWIISEVCW-UHFFFAOYSA-N | |
butylboronic acid | |
20479 | |
98% |
92°C to 96°C | |
MFCD00002106 | |
Air sensitive; Hygroscopic | |
Soluble in water. | |
CCCCB(O)O | |
101.94 | |
101.94 | |
1-Butylboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 224-607-7
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.