Learn More
1-Bromoperfluorooctane, 98+%, Thermo Scientific Chemicals
22.60 € - 235.00 €
Chemical Identifiers
CAS | 423-55-2 |
---|---|
Molecular Formula | C8BrF17 |
Molecular Weight (g/mol) | 498.97 |
MDL Number | MFCD00042082 |
InChI Key | WTWWXOGTJWMJHI-UHFFFAOYSA-N |
Synonym | perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide |
PubChem CID | 9873 |
ChEBI | CHEBI:38803 |
IUPAC Name | 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane |
SMILES | FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410647
|
Thermo Scientific Alfa Aesar
B22470.03 |
1 g | N/A | ||||||
11420647
|
Thermo Scientific Alfa Aesar
B22470.06 |
5 g |
67.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430647
|
Thermo Scientific Alfa Aesar
B22470.14 |
25 g |
235.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromoperfluorooctane 1-Bromoheptadecafluorooctane (perfluorooctyl bromide) can be used as contrast agent in the vascular system for fluorine-19 magnetic resonance imaging or as synthetic oxygen carrier. It is used as an O2 carrier in some emulsions.
Solubility
Not miscible or difficult to mix in water.
Notes
Incompatible with oxidizing agents, acids, metals.
Chemical Identifiers
423-55-2 | |
498.97 | |
WTWWXOGTJWMJHI-UHFFFAOYSA-N | |
9873 | |
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane |
C8BrF17 | |
MFCD00042082 | |
perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide | |
CHEBI:38803 | |
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br |
Specifications
423-55-2 | |
1.93 | |
None | |
CF3(CF2)7Br | |
MFCD00042082 | |
14,7156 | |
Not miscible or difficult to mix in water. | |
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br | |
498.97 | |
CHEBI:38803 | |
≥98% |
6°C | |
141°C to 143°C | |
C8BrF17 | |
1.305 | |
2016022 | |
perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide | |
WTWWXOGTJWMJHI-UHFFFAOYSA-N | |
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane | |
9873 | |
498.98 | |
1-Bromoperfluorooctane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 207-028-4
missing translation for 'rtecsNumber' : RG8700000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only