missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctane, 98+%, Thermo Scientific Chemicals

Product Code. 11407756
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-83-1
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.13
MDL Number MFCD00000276
InChI Key VMKOFRJSULQZRM-UHFFFAOYSA-N
Synonym n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
PubChem CID 8140
IUPAC Name 1-bromooctane
SMILES CCCCCCCCBr

Specifications

Melting Point -55°C
Density 1.114
Boiling Point 200°C to 202°C
Flash Point 78°C (172°F)
Odor Characteristic
Linear Formula CH3(CH2)7Br
Refractive Index 1.4518
Quantity 1000 g
UN Number UN3082
Beilstein 1733136
Merck Index 14,6764
Solubility Information Immiscible with water.
Formula Weight 193.13
Percent Purity ≥98%
Chemical Name or Material 1-Bromooctane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'