Learn More
1-Bromooctadecane, 97%, Thermo Scientific Chemicals
25.33 € - 234.17 €
Chemical Identifiers
CAS | 112-89-0 |
---|---|
Molecular Formula | C18H37Br |
Molecular Weight (g/mol) | 333.40 |
MDL Number | MFCD00000231 |
InChI Key | WSULSMOGMLRGKU-UHFFFAOYSA-N |
Synonym | octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane |
PubChem CID | 8218 |
IUPAC Name | 1-bromooctadecane |
SMILES | CCCCCCCCCCCCCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11487427
|
Thermo Scientific Alfa Aesar
A15588.22 |
100 g |
25.33 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11497427
|
Thermo Scientific Alfa Aesar
A15588.36 |
500 g |
63.11 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11407437
|
Thermo Scientific Alfa Aesar
A15588.0E |
2500 g |
234.17 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.
Solubility
Soluble in acetone and most common organic solvents. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
112-89-0 | |
333.40 | |
WSULSMOGMLRGKU-UHFFFAOYSA-N | |
8218 | |
CCCCCCCCCCCCCCCCCCBr |
C18H37Br | |
MFCD00000231 | |
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane | |
1-bromooctadecane |
Specifications
112-89-0 | |
0.976 | |
>110°C (230°F) | |
C18H37Br | |
MFCD00000231 | |
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane | |
WSULSMOGMLRGKU-UHFFFAOYSA-N | |
1-bromooctadecane | |
8218 | |
97% |
25°C to 30°C | |
150°C to 152°C (0.1 mmHg) | |
Odorless | |
CH3(CH2)17Br | |
774145 | |
Soluble in acetone and most common organic solvents. Insoluble in water. | |
CCCCCCCCCCCCCCCCCCBr | |
333.40 | |
333.39 | |
1-Bromooctadecane |
Safety and Handling
missing translation for 'einecsNumber' : 204-013-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.