missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals

Product Code. 15481169
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as a intermediate for organic synthesis and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3163-27-7
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.097
MDL Number MFCD00010804
InChI Key RZJGKPNCYQZFGR-UHFFFAOYSA-N
Synonym 1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389
PubChem CID 137844
IUPAC Name 1-(bromomethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CBr

Specifications

Color White
Density 1.44 g/mL
Boiling Point 213°C (100 mmHg)
Refractive Index 1.66
Quantity 100 g
UN Number UN3261
Beilstein 742417
Solubility Information Slightly soluble in water.
Formula Weight 221.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-(Bromomethyl)naphthalene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.