Learn More
1-(Bromomethyl)cyclopropane, 95%, Thermo Scientific™
21.00 € - 298.10 €
Chemical Identifiers
CAS | 7051-34-5 |
---|---|
Molecular Formula | C4H7Br |
Molecular Weight (g/mol) | 135.00 |
MDL Number | MFCD00001306 |
InChI Key | AEILLAXRDHDKDY-UHFFFAOYSA-N |
Synonym | bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide |
PubChem CID | 81503 |
SMILES | BrCC1CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10205904
|
Thermo Scientific Maybridge
RJF00467DA |
1 g | Amber Glass Bottle | N/A | |||||
10080062
|
Thermo Scientific Maybridge
RJF00467EA |
10 g | Amber Glass Bottle | N/A | |||||
10258242
|
Thermo Scientific Maybridge
RJF00467EB |
25 g | Amber Glass Bottle | N/A | |||||
Description
Chemical Identifiers
7051-34-5 | |
135.00 | |
AEILLAXRDHDKDY-UHFFFAOYSA-N | |
81503 |
C4H7Br | |
MFCD00001306 | |
bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide | |
BrCC1CC1 |
Specifications
7051-34-5 | |
1.3800g/mL | |
C4H7Br | |
MFCD00001306 | |
AEILLAXRDHDKDY-UHFFFAOYSA-N | |
(bromomethyl)cyclopropane | |
81503 | |
95% |
95.0 | |
95% | |
C3H5CH2Br | |
bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide | |
BrCC1CC1 | |
135.00 | |
135 | |
1-(Bromomethyl)cyclopropane |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.