missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)adamantane, 97%, Thermo Scientific™

Product Code. 10396494
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 14651-42-4
Molecular Formula C11H17Br
Molecular Weight (g/mol) 229.161
InChI Key RLRBYCAVZBQANU-UHFFFAOYSA-N
Synonym 1-bromomethyl adamantane, tricyclo 3.3.1.13,7 decane, 1-bromomethyl, adamantanylbromomethane, 1-bromomethyl tricyclo 3.3.1.1∼3,7∼ decane, 3r,5s,7s-1-bromomethyl adamantane, pubchem21524, 1-adamantylmethylbromide, 1-bromoethyl adamantane, 1-adamantyl methyl bromide, 1s,3r,5s,7s-1-bromomethyl adamantane
PubChem CID 1416304
IUPAC Name 1-(bromomethyl)adamantane
SMILES C1C2CC3CC1CC(C2)(C3)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 229.16
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)adamantane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.