Learn More
1-Bromoheptane, 99%, Thermo Scientific Chemicals
20.90 € - 168.00 €
Chemical Identifiers
CAS | 629-04-9 |
---|---|
Molecular Formula | C7H15Br |
Molecular Weight (g/mol) | 179.10 |
MDL Number | MFCD00000273 |
InChI Key | LSXKDWGTSHCFPP-UHFFFAOYSA-N |
Synonym | heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane |
PubChem CID | 12369 |
IUPAC Name | 1-bromoheptane |
SMILES | CCCCCCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10020680
|
Thermo Scientific Acros
107030050 |
5 mL | Glass bottle | ||||||
10467670
|
Thermo Scientific Acros
107032500 |
250 mL | Glass bottle |
47.70 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10656754
|
Thermo Scientific Acros
107030010 |
1 L | Glass bottle |
168.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
629-04-9 | |
179.10 | |
LSXKDWGTSHCFPP-UHFFFAOYSA-N | |
12369 | |
CCCCCCCBr |
C7H15Br | |
MFCD00000273 | |
heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane | |
1-bromoheptane |
Specifications
629-04-9 | |
100.0 | |
Colorless to Yellow | |
180°C | |
Authentic | |
C7H15Br | |
1.4489 to 1.4509 | |
01, 155 | |
heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane | |
LSXKDWGTSHCFPP-UHFFFAOYSA-N | |
1-bromoheptane | |
12369 | |
99% | |
1-Bromoheptane, 99% |
98.5 | |
-58°C | |
1.1400g/mL | |
60°C | |
99% | |
CH3(CH2)6Br | |
MFCD00000273 | |
1.14 | |
Solubility in water: insoluble | |
CCCCCCCBr | |
179.10 | |
179.1 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 211-068-8
missing translation for 'rtecsNumber' : MI8100000
missing translation for 'tsca' : TSCA
RUO – Research Use Only