missing translation for 'onlineSavingsMsg'
Learn More

(1-Bromoethyl)benzene, 97%, Thermo Scientific Chemicals

Product Code. 11422868
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(1-Bromoethyl)benzene has been employed in controlled radical polymerization of styrene, in asymmetric esterification of benzoic acid in the presence of a chiral cyclic guanidine and as initiator in the synthesis of bromine terminated polyp-methoxystyrene and polystyrene via atom transfer radical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Bromoethyl)benzene has been employed in controlled radical polymerization of styrene, in asymmetric esterification of benzoic acid in the presence of a chiral cyclic guanidine and as initiator in the synthesis of bromine terminated polyp-methoxystyrene and polystyrene via atom transfer radical polymerization.

Solubility
Soluble in alcohol, ether, and benzene.

Notes
Moisture Sensitive. Do not store together with strongly basic and oxidizing materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-71-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000139
InChI Key CRRUGYDDEMGVDY-UHFFFAOYSA-N
Synonym 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
PubChem CID 11454
IUPAC Name 1-bromoethylbenzene
SMILES CC(C1=CC=CC=C1)Br

Specifications

Melting Point -65°C
Density 1.356
Boiling Point 200°C
Flash Point 81°C (177°F)
Refractive Index 1.56
Quantity 100 g
Beilstein 507210
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohol,ether,and benzene.
Formula Weight 185.06
Percent Purity 97%
Chemical Name or Material (1-Bromoethyl)benzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.