Learn More
1-Bromododecane, 98%, Thermo Scientific Chemicals
38.39 € - 464.31 €
Chemical Identifiers
CAS | 143-15-7 |
---|---|
Molecular Formula | C12H25Br |
Molecular Weight (g/mol) | 249.236 |
MDL Number | MFCD00000225 |
InChI Key | PBLNBZIONSLZBU-UHFFFAOYSA-N |
Synonym | dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d |
PubChem CID | 8919 |
IUPAC Name | 1-bromododecane |
SMILES | CCCCCCCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413284
|
Thermo Scientific Alfa Aesar
A10600.30 |
250 g |
38.39 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403284
|
Thermo Scientific Alfa Aesar
A10600.0B |
1000 g |
108.54 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423284
|
Thermo Scientific Alfa Aesar
A10600.0I |
5000 g |
464.31 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
143-15-7 | |
249.236 | |
PBLNBZIONSLZBU-UHFFFAOYSA-N | |
8919 | |
CCCCCCCCCCCCBr |
C12H25Br | |
MFCD00000225 | |
dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d | |
1-bromododecane |
Specifications
143-15-7 | |
1.04 | |
146°C (294°F) | |
C12H25Br | |
1.458 | |
506159 | |
dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d | |
PBLNBZIONSLZBU-UHFFFAOYSA-N | |
1-bromododecane | |
8919 | |
98% |
-10°C | |
276°C | |
Weak | |
CH3(CH2)11Br | |
MFCD00000225 | |
14,5389 | |
Insoluble in water. | |
CCCCCCCCCCCCBr | |
249.236 | |
249.24 | |
1-Bromododecane |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 205-587-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1-Bromododecane
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Lauryl bromide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.