Learn More
1-Bromobutane, 99%, Thermo Scientific Chemicals
27.34 € - 225.12 €
Chemical Identifiers
CAS | 109-65-9 |
---|---|
Molecular Formula | C4H9Br |
Molecular Weight (g/mol) | 137.02 |
MDL Number | MFCD00000260 |
InChI Key | MPPPKRYCTPRNTB-UHFFFAOYSA-N |
Synonym | butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831 |
PubChem CID | 8002 |
IUPAC Name | 1-bromobutane |
SMILES | CCCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10387763
|
Thermo Scientific Acros
106770500 |
50 mL | Glass bottle |
27.34 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10509820
|
Thermo Scientific Acros
106772500 |
250 mL | Glass bottle |
39.09 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10704601
|
Thermo Scientific Acros
106770010 |
1 L | Glass bottle |
106.53 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10338853
|
Thermo Scientific Acros
106770025 |
2.5 L | Glass bottle |
225.12 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-65-9 | |
137.02 | |
MPPPKRYCTPRNTB-UHFFFAOYSA-N | |
8002 | |
CCCCBr |
C4H9Br | |
MFCD00000260 | |
butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831 | |
1-bromobutane |
Specifications
109-65-9 | |
100.0 | |
Colorless to Yellow | |
100.0°C to 104.0°C | |
Authentic | |
C4H9Br | |
1.4380 to 1.4404 | |
01,119 | |
1.27 | |
Solubility in water: 0.608g/L (30°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene and ccl4 | |
CCCCBr | |
137.02 | |
137.02 | |
Liquid |
98.5 | |
-112.0°C | |
1.2700g/mL | |
10°C | |
98.5% min. (GC) | |
CH3(CH2)3Br | |
MFCD00000260 | |
15,1555 | |
butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831 | |
MPPPKRYCTPRNTB-UHFFFAOYSA-N | |
1-bromobutane | |
8002 | |
99% | |
1-Bromobutane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-691-9
- 1-Bromobutane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.