missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-iodobenzene, 98%, Thermo Scientific Chemicals
99.60 € - 285.42 €
Chemical Identifiers
CAS | 589-87-7 |
---|---|
Molecular Formula | C6H4BrI |
Molecular Weight (g/mol) | 282.9 |
InChI Key | UCCUXODGPMAHRL-UHFFFAOYSA-N |
Synonym | 4-bromoiodobenzene, benzene, 1-bromo-4-iodo, p-bromoiodobenzene, p-iodobromobenzene, p-bromophenyl iodide, 4-bromo-1-iodobenzene, 1-bromo-4-iodo-benzene, 4-iodobromobenzene, rarechem fh 1g 0a12, p-bromo iodobenzene |
PubChem CID | 11522 |
IUPAC Name | 1-bromo-4-iodobenzene |
SMILES | C1=CC(=CC=C1Br)I |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10553961
|
Thermo Scientific
218380250 |
25 g | Glass bottle |
99.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964521
|
Thermo Scientific Acros
218381000 |
100 g | Glass bottle |
285.42 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
589-87-7 | |
282.9 | |
4-bromoiodobenzene, benzene, 1-bromo-4-iodo, p-bromoiodobenzene, p-iodobromobenzene, p-bromophenyl iodide, 4-bromo-1-iodobenzene, 1-bromo-4-iodo-benzene, 4-iodobromobenzene, rarechem fh 1g 0a12, p-bromo iodobenzene | |
1-bromo-4-iodobenzene |
C6H4BrI | |
UCCUXODGPMAHRL-UHFFFAOYSA-N | |
11522 | |
C1=CC(=CC=C1Br)I |
Specifications
589-87-7 | |
White to Brown | |
Authentic | |
C6H4BrI | |
05,223 | |
UCCUXODGPMAHRL-UHFFFAOYSA-N | |
1-bromo-4-iodobenzene | |
11522 | |
98% | |
1-Bromo-4-iodobenzene |
89.0°C to 93.0°C | |
120.0°C to 122.0°C (14.0 mmHg) | |
97.5% min. (GC) | |
BrC6H4I | |
4-bromoiodobenzene, benzene, 1-bromo-4-iodo, p-bromoiodobenzene, p-iodobromobenzene, p-bromophenyl iodide, 4-bromo-1-iodobenzene, 1-bromo-4-iodo-benzene, 4-iodobromobenzene, rarechem fh 1g 0a12, p-bromo iodobenzene | |
C1=CC(=CC=C1Br)I | |
282.9 | |
282.9 | |
Crystalline Powder, Crystals or Needles |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-662-7
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Bromo-4-iodobenzene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title