Learn More
1-Bromo-3-(tert-butyldimethylsiloxy)benzene, 98+%, Thermo Scientific Chemicals
28.40 € - 94.00 €
Chemical Identifiers
CAS | 65423-56-5 |
---|---|
Molecular Formula | C12H19BrOSi |
Molecular Weight (g/mol) | 287.272 |
MDL Number | MFCD01318106 |
InChI Key | BFRHMVJJJGUHDI-UHFFFAOYSA-N |
Synonym | 3-bromophenoxy tert-butyl dimethylsilane, 3-bromophenyl tert-butyldimethylsilyl ether, 1-bromo-3-tert-butyldimethylsiloxy benzene, silane, 3-bromophenoxy 1,1-dimethylethyl dimethyl, acmc-1b3kt, 3-t-butyldimethylsilyloxybromobenzene, 3-t-butyldimethylsilyloxy bromobenzene, 3-bromophenoxy-tert-butyldimethylsilane, 3-tert-butyldimethylsiloxy bromobenzene |
PubChem CID | 11254705 |
IUPAC Name | (3-bromophenoxy)-tert-butyl-dimethylsilane |
SMILES | CC(C)(C)[Si](C)(C)OC1=CC(=CC=C1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348406
|
Thermo Scientific Alfa Aesar
L17440.03 |
1 g | N/A | ||||||
11358406
|
Thermo Scientific Alfa Aesar
L17440.06 |
5 g |
94.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
65423-56-5 | |
287.272 | |
BFRHMVJJJGUHDI-UHFFFAOYSA-N | |
11254705 | |
CC(C)(C)[Si](C)(C)OC1=CC(=CC=C1)Br |
C12H19BrOSi | |
MFCD01318106 | |
3-bromophenoxy tert-butyl dimethylsilane, 3-bromophenyl tert-butyldimethylsilyl ether, 1-bromo-3-tert-butyldimethylsiloxy benzene, silane, 3-bromophenoxy 1,1-dimethylethyl dimethyl, acmc-1b3kt, 3-t-butyldimethylsilyloxybromobenzene, 3-t-butyldimethylsilyloxy bromobenzene, 3-bromophenoxy-tert-butyldimethylsilane, 3-tert-butyldimethylsiloxy bromobenzene | |
(3-bromophenoxy)-tert-butyl-dimethylsilane |
Specifications
66°C (0.3mmHg) | |
C12H19BrOSi | |
2256298 | |
BFRHMVJJJGUHDI-UHFFFAOYSA-N | |
(3-bromophenoxy)-tert-butyl-dimethylsilane | |
11254705 | |
≥98% | |
Moisture sensitive |
65423-56-5 | |
MFCD01318106 | |
3-bromophenoxy tert-butyl dimethylsilane, 3-bromophenyl tert-butyldimethylsilyl ether, 1-bromo-3-tert-butyldimethylsiloxy benzene, silane, 3-bromophenoxy 1,1-dimethylethyl dimethyl, acmc-1b3kt, 3-t-butyldimethylsilyloxybromobenzene, 3-t-butyldimethylsilyloxy bromobenzene, 3-bromophenoxy-tert-butyldimethylsilane, 3-tert-butyldimethylsiloxy bromobenzene | |
CC(C)(C)[Si](C)(C)OC1=CC(=CC=C1)Br | |
287.272 | |
287.27 | |
1.509 | |
1-Bromo-3-(tert-butyldimethylsiloxy)benzene |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.