Learn More
1-Bromo-3-methyl-2-butene, 90+%, stab. with silver, Thermo Scientific Chemicals
58.19 € - 859.28 €
Chemical Identifiers
CAS | 870-63-3 |
---|---|
Molecular Formula | C5H9Br |
Molecular Weight (g/mol) | 149.031 |
MDL Number | MFCD00000242 |
InChI Key | LOYZVRIHVZEDMW-UHFFFAOYSA-N |
Synonym | 1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide |
PubChem CID | 70092 |
IUPAC Name | 1-bromo-3-methylbut-2-ene |
SMILES | CC(=CCBr)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435383
|
Thermo Scientific Alfa Aesar
A13388.09 |
10 g |
58.19 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445383
|
Thermo Scientific Alfa Aesar
A13388.18 |
50 g |
211.05 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455383
|
Thermo Scientific Alfa Aesar
A13388.30 |
250 g |
859.28 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromo-3-methyl-2-butene is considered as a source of 3,3-dimethylallyl group and used in the preparation of natural products. It is also employed in the synthesis of base-modified pyrimidine nucleosides. Further, it is used in the preparation of (±)-eldanolide, 1-(3,3-dimethylallyl)-L-tryptophan and (±)-fumagillin, an antibiotic isolated from Aspergillus fumigattus.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-3-methyl-2-butene is considered as a source of 3,3-dimethylallyl group and used in the preparation of natural products. It is also employed in the synthesis of base-modified pyrimidine nucleosides. Further, it is used in the preparation of (±)-eldanolide, 1-(3,3-dimethylallyl)-L-tryptophan and (±)-fumagillin, an antibiotic isolated from Aspergillus fumigattus.
Solubility
Immiscible with water
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases
Chemical Identifiers
870-63-3 | |
149.031 | |
LOYZVRIHVZEDMW-UHFFFAOYSA-N | |
70092 | |
CC(=CCBr)C |
C5H9Br | |
MFCD00000242 | |
1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide | |
1-bromo-3-methylbut-2-ene |
Specifications
870-63-3 | |
135°C | |
Pungent | |
1.4895 | |
UN2920 | |
Light sensitive | |
Immiscible with water | |
CC(=CCBr)C | |
149.031 | |
149.03 | |
1-Bromo-3-methyl-2-butene, stabilized with silver |
1.281 | |
32°C (90°F) | |
C5H9Br | |
MFCD00000242 | |
1420778 | |
1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide | |
LOYZVRIHVZEDMW-UHFFFAOYSA-N | |
1-bromo-3-methylbut-2-ene | |
70092 | |
≥90% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P374-P380-P501c
H226-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 212-799-5
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.