missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-fluoro-5-nitrobenzene, 98%, Thermo Scientific™

Product Code. 15467698
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7087-65-2
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 219.997
MDL Number MFCD09955454
InChI Key SWXVEPMSQBEVRH-UHFFFAOYSA-N
Synonym 3-bromo-5-fluoronitrobenzene, 1-nitro-3-fluoro-5-bromobenzene, benzene, 1-bromo-3-fluoro-5-nitro, pubchem17569, acmc-209ogb, 3-fluoro-5-nitrobromobenzene, 5-bromo-3-fluoro-1-nitrobenzene
PubChem CID 15020157
IUPAC Name 1-bromo-3-fluoro-5-nitrobenzene
SMILES C1=C(C=C(C=C1F)Br)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 5 g
UN Number UN2810
Formula Weight 220
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-fluoro-5-nitrobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'