Learn More
1-Boc-indole, 97%, Thermo Scientific Chemicals
49.45 € - 171.86 €
Chemical Identifiers
CAS | 75400-67-8 |
---|---|
Molecular Formula | C13H15NO2 |
Molecular Weight (g/mol) | 217.27 |
MDL Number | MFCD02093939 |
InChI Key | OWPIFQXNMLDXKW-UHFFFAOYSA-N |
Synonym | 1-boc-indole, tert-butyl 1h-indole-1-carboxylate, n-boc-indole, tert-butyl 1-indolecarboxylate, 1h-indole-1-carboxylic acid, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl indole, n-boc indole, pubchem7228, acmc-1bjgd, 1-tert-butoxycarbonylindole |
PubChem CID | 3532980 |
SMILES | CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358546
|
Thermo Scientific Alfa Aesar
L17601.03 |
1 g |
49.45 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368546
|
Thermo Scientific Alfa Aesar
L17601.06 |
5 g |
171.86 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75400-67-8 | |
217.27 | |
OWPIFQXNMLDXKW-UHFFFAOYSA-N | |
3532980 |
C13H15NO2 | |
MFCD02093939 | |
1-boc-indole, tert-butyl 1h-indole-1-carboxylate, n-boc-indole, tert-butyl 1-indolecarboxylate, 1h-indole-1-carboxylic acid, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl indole, n-boc indole, pubchem7228, acmc-1bjgd, 1-tert-butoxycarbonylindole | |
CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12 |
Specifications
75400-67-8 | |
113°C to 114°C (0.2 mmHg) | |
1.543 | |
4674239 | |
OWPIFQXNMLDXKW-UHFFFAOYSA-N | |
tert-butyl 1H-indole-1-carboxylate | |
3532980 | |
97% |
1.07 | |
C13H15NO2 | |
MFCD02093939 | |
1-boc-indole, tert-butyl 1h-indole-1-carboxylate, n-boc-indole, tert-butyl 1-indolecarboxylate, 1h-indole-1-carboxylic acid, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl indole, n-boc indole, pubchem7228, acmc-1bjgd, 1-tert-butoxycarbonylindole | |
CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12 | |
217.27 | |
217.27 | |
1-Boc-indole |
Safety and Handling
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.