missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoyl-4-piperidone, 98+%, Thermo Scientific Chemicals

Product Code. 11481608
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24686-78-0
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.24
MDL Number MFCD00006189
InChI Key NZAXGZYPZGEVBD-UHFFFAOYSA-N
Synonym n-benzoyl-4-piperidone, 1-benzoyl-4-piperidinone, 4-piperidinone, 1-benzoyl, 1-benzoyl-4-piperidone, 1-benzoyl-4-oxopiperidine, 4-piperidone, 1-benzoyl, 1-phenylcarbonyl piperidin-4-one, 1-benzoyl-piperidin-4-one, n-benzoyl-4piperidone, 1-benzoyl-piperidinoine
PubChem CID 90583
IUPAC Name 1-benzoylpiperidin-4-one
SMILES O=C(N1CCC(=O)CC1)C1=CC=CC=C1

Specifications

Melting Point 52°C to 57°C
Boiling Point 158°C to 160°C (0.2 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 158848
Formula Weight 203.24
Percent Purity ≥98%
Chemical Name or Material 1-Benzoyl-4-piperidone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'