Learn More
1-Amino-1-cyclopropanecarbonitrile hydrochloride, 97%, Thermo Scientific Chemicals
27.00 € - 248.00 €
Chemical Identifiers
CAS | 127946-77-4 |
---|---|
Molecular Formula | C4H7ClN2 |
Molecular Weight (g/mol) | 118.564 |
MDL Number | MFCD04114063 |
InChI Key | PCEIEQLJYDMRFZ-UHFFFAOYSA-N |
Synonym | 1-amino-1-cyclopropanecarbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hydrochloride, 1-aminocyclopropane-1-carbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hcl, 1-amino-cyclopropyl cyanic hydrochloride, 1-amino-1-cyanocyclopropane hydrochloride, 1-amino-cyclopropanecarbonitrile hydrochloride, cyclopropanecarbonitrile, 1-amino-, monohydrochloride, 1-cyanocyclopropanaminium chloride, pubchem16832 |
PubChem CID | 14586363 |
IUPAC Name | 1-aminocyclopropane-1-carbonitrile;hydrochloride |
SMILES | C1CC1(C#N)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15447675
|
Thermo Scientific Alfa Aesar
H27288.MD |
250 mg | |||||||
11312349
|
Thermo Scientific Alfa Aesar
H27288.03 |
1 g |
77.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11322349
|
Thermo Scientific Alfa Aesar
H27288.06 |
5 g |
248.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
127946-77-4 | |
118.564 | |
PCEIEQLJYDMRFZ-UHFFFAOYSA-N | |
14586363 | |
C1CC1(C#N)N.Cl |
C4H7ClN2 | |
MFCD04114063 | |
1-amino-1-cyclopropanecarbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hydrochloride, 1-aminocyclopropane-1-carbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hcl, 1-amino-cyclopropyl cyanic hydrochloride, 1-amino-1-cyanocyclopropane hydrochloride, 1-amino-cyclopropanecarbonitrile hydrochloride, cyclopropanecarbonitrile, 1-amino-, monohydrochloride, 1-cyanocyclopropanaminium chloride, pubchem16832 | |
1-aminocyclopropane-1-carbonitrile;hydrochloride |
Specifications
127946-77-4 | |
C4H7ClN2 | |
UN3439 | |
PCEIEQLJYDMRFZ-UHFFFAOYSA-N | |
1-aminocyclopropane-1-carbonitrile;hydrochloride | |
14586363 | |
97% |
∽220°C (decomposition) | |
MFCD04114063 | |
1-amino-1-cyclopropanecarbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hydrochloride, 1-aminocyclopropane-1-carbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hcl, 1-amino-cyclopropyl cyanic hydrochloride, 1-amino-1-cyanocyclopropane hydrochloride, 1-amino-cyclopropanecarbonitrile hydrochloride, cyclopropanecarbonitrile, 1-amino-, monohydrochloride, 1-cyanocyclopropanaminium chloride, pubchem16832 | |
C1CC1(C#N)N.Cl | |
118.564 | |
118.56 | |
1-Amino-1-cyclopropanecarbonitrile hydrochloride |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only