missing translation for 'onlineSavingsMsg'
Learn More

1-Allylcyclohexanol, 97%, Thermo Scientific Chemicals

Product Code. 11459366
Click to view available options
Quantity:
2.5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-34-8
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00021403
InChI Key ZSLKGUQYEQVKQE-UHFFFAOYSA-N
Synonym 1-allylcyclohexanol, cyclohexanol, 1-allyl, 1-allyl-1-cyclohexanol, cyclohexanol, 1-2-propenyl, 1-prop-2-en-1-yl cyclohexan-1-ol, allylcyclohexanol, 1-allyl-cyclohexan-1-ol, acmc-1bt3v, 4-06-00-00235 beilstein handbook reference, 1-prop-2-enyl-1-cyclohexanol
PubChem CID 79145
IUPAC Name 1-prop-2-enylcyclohexan-1-ol
SMILES C=CCC1(CCCCC1)O

Specifications

Quantity 2.5 g
Formula Weight 140.22
Percent Purity 97%
Chemical Name or Material 1-Allylcyclohexanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'