Learn More
1,9-Nonanedithiol, 95%, Thermo Scientific Chemicals
50.40 € - 158.00 €
Chemical Identifiers
CAS | 3489-28-9 |
---|---|
Molecular Formula | C9H20S2 |
Molecular Weight (g/mol) | 192.38 |
MDL Number | MFCD00004914 |
InChI Key | GJRCLMJHPWCJEI-UHFFFAOYSA-N |
Synonym | 1,9-nonanedithiol, 1,9-dimercaptononane, nonamethylene dimercaptan, unii-5bpj1xqe3t, 5bpj1xqe3t, fema no. 3513, 1,9-nonane dithiol, 1,9-dinonyl mercaptan, acmc-20ap0g |
PubChem CID | 248488 |
IUPAC Name | nonane-1,9-dithiol |
SMILES | SCCCCCCCCCS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327166
|
Thermo Scientific Alfa Aesar
L07172.03 |
1 g |
50.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337166
|
Thermo Scientific Alfa Aesar
L07172.06 |
5 g |
158.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3489-28-9 | |
192.38 | |
GJRCLMJHPWCJEI-UHFFFAOYSA-N | |
248488 | |
SCCCCCCCCCS |
C9H20S2 | |
MFCD00004914 | |
1,9-nonanedithiol, 1,9-dimercaptononane, nonamethylene dimercaptan, unii-5bpj1xqe3t, 5bpj1xqe3t, fema no. 3513, 1,9-nonane dithiol, 1,9-dinonyl mercaptan, acmc-20ap0g | |
nonane-1,9-dithiol |
Specifications
3489-28-9 | |
351°C to 352°C | |
C9H20S2 | |
MFCD00004914 | |
Air Sensitive | |
GJRCLMJHPWCJEI-UHFFFAOYSA-N | |
nonane-1,9-dithiol | |
248488 | |
97% |
0.953 | |
>100°C (212°F) | |
1.4999 | |
1737568 | |
1,9-nonanedithiol, 1,9-dimercaptononane, nonamethylene dimercaptan, unii-5bpj1xqe3t, 5bpj1xqe3t, fema no. 3513, 1,9-nonane dithiol, 1,9-dinonyl mercaptan, acmc-20ap0g | |
SCCCCCCCCCS | |
192.38 | |
192.39 | |
1,9-Nonanedithiol |
Safety and Handling
GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 222-482-3
missing translation for 'tsca' : Yes
Recommended Storage : Store under Argon
RUO – Research Use Only