missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dibromononane, 97%, Thermo Scientific Chemicals

Product Code. 11424210
Click to view available options
Quantity:
10 g
25 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-33-1
Molecular Formula C9H18Br2
Molecular Weight (g/mol) 286.051
MDL Number MFCD00000279
InChI Key WGAXVZXBFBHLMC-UHFFFAOYSA-N
Synonym nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
PubChem CID 20677
IUPAC Name 1,9-dibromononane
SMILES C(CCCCBr)CCCCBr

Specifications

Density 1.41
Boiling Point 285°C to 288°C
Flash Point >110°C (230°F)
Linear Formula Br(CH2)9Br
Refractive Index 1.4965
Quantity 25 g
Beilstein 1737598
Formula Weight 286.06
Percent Purity 97%
Chemical Name or Material 1,9-Dibromononane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'