missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibromooctane, 98%, Thermo Scientific Chemicals

Product Code. 11437567
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,8-Dibromooctane is used as an additive in the preparation poly[[4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b']dithiophene-2,6-diyl][3-fluoro-2-[(2-ethylhexyl)carbonyl]thieno[3,4-b]thiophenediyl]]:[6,6]-phenylC71-butyric acid methyl ester, which finds application in the fabrication of organic photovoltaics (OPVs). Further, it is used in the preparation of (1,4-bis-(8-bromooctyloxy)-benzene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Dibromooctane is used as an additive in the preparation poly[[4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl][3-fluoro-2-[(2-ethylhexyl)carbonyl]thieno[3,4-b]thiophenediyl]]:[6,6]-phenylC71-butyric acid methyl ester, which finds application in the fabrication of organic photovoltaics (OPVs). Further, it is used in the preparation of (1,4-bis-(8-bromooctyloxy)-benzene).

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-32-0
Molecular Formula C8H16Br2
MDL Number MFCD00000277
InChI Key DKEGCUDAFWNSSO-UHFFFAOYSA-N
Synonym octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r
PubChem CID 78310
IUPAC Name 1,8-dibromooctane

Specifications

Melting Point 15°C to 16°C
Density 1.468
Boiling Point 270°C to 272°C
Flash Point >110°C (230°F)
Refractive Index 1.4995
Quantity 50 g
Beilstein 1698114
Solubility Information Immiscible with water.
SMILES C(CCCCBr)CCCBr
Molecular Weight (g/mol) 272.024
Formula Weight 272.02
Percent Purity 98%
Chemical Name or Material 1,8-Dibromooctane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'