Learn More
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%, Thermo Scientific Chemicals
11.20 € - 2014.02 €
Chemical Identifiers
CAS | 6674-22-2 |
---|---|
MDL Number | MFCD00006930 |
InChI Key | GQHTUMJGOHRCHB-UHFFFAOYSA-N |
Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene |
PubChem CID | 81184 |
IUPAC Name | 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine |
SMILES | C1CCC2=NCCCN2CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011970
|
Thermo Scientific Acros
160610050 |
5 g | Glass bottle | N/A | |||||
10104320
|
Thermo Scientific Acros
160610250 |
25 g | Glass bottle |
44.62 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10377240
|
Thermo Scientific Acros
160611000 |
100 g | Glass bottle |
123.62 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10662023
|
Thermo Scientific Acros
160615000 |
500 g | Glass bottle |
449.24 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10023313
|
Thermo Scientific Acros
160610025 |
2.5 kg | Glass bottle |
2014.02 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6674-22-2 | |
GQHTUMJGOHRCHB-UHFFFAOYSA-N | |
81184 | |
C1CCC2=NCCCN2CC1 |
MFCD00006930 | |
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene | |
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine |
Specifications
6674-22-2 | |
100.0 | |
1.019g/mL | |
98% | |
1.5210 to 1.5230 (20°C, 589nm) | |
23, V,5, 271 | |
15, 2996 | |
GQHTUMJGOHRCHB-UHFFFAOYSA-N | |
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine | |
81184 | |
≥97.5% | |
1, 8-Diazabicyclo[5.4.0]undec-7-ene, 98% |
97.5 | |
Colorless to Yellow | |
Authentic | |
C9H16N2 | |
MFCD00006930 | |
02,101; 04,119; 05,177; 06,158; 07,87; 11,155; 13,91; 14,109; 15,109; 16,105; 17,99 | |
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene | |
C1CCC2=NCCCN2CC1 | |
152.241 | |
152.24 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 229-713-7
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.