Learn More
1,5-Dibromopentane, 98%, Thermo Scientific Chemicals
32.66 € - 410.04 €
Chemical Identifiers
CAS | 111-24-0 |
---|---|
Molecular Formula | C5H10Br2 |
Molecular Weight (g/mol) | 229.943 |
MDL Number | MFCD00000268 |
InChI Key | IBODDUNKEPPBKW-UHFFFAOYSA-N |
PubChem CID | 8100 |
IUPAC Name | 1,5-dibromopentane |
SMILES | C(CCBr)CCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11438387
|
Thermo Scientific Alfa Aesar
A18465.22 |
100 g |
32.66 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448387
|
Thermo Scientific Alfa Aesar
A18465.36 |
500 g |
104.52 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458387
|
Thermo Scientific Alfa Aesar
A18465.0E |
2500 g |
410.04 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.
Solubility
Miscible with water, slightly miscible with chloroform and benzene.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
111-24-0 | |
229.943 | |
IBODDUNKEPPBKW-UHFFFAOYSA-N | |
1,5-dibromopentane |
C5H10Br2 | |
MFCD00000268 | |
8100 | |
C(CCBr)CCBr |
Specifications
111-24-0 | |
1.688 | |
79°C (174°F) | |
1.5122 | |
1209245 | |
Miscible with water,slightly miscible with chloroform and benzene. | |
C(CCBr)CCBr | |
229.943 | |
229.95 | |
1,5-Dibromopentane |
-34°C | |
224°C | |
C5H10Br2 | |
MFCD00000268 | |
pentamethylene bromide, pentane, 1,5-dibromo, pentamethylene dibromide, unii-b8q228qys1, pentane-1,5-dibromide, 1,5-dibromo-pentane, 1,5-pentane dibromide, pentane,1,5-dibromo, br ch2 5br, ccris 8918 | |
IBODDUNKEPPBKW-UHFFFAOYSA-N | |
1,5-dibromopentane | |
8100 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319
missing translation for 'einecsNumber' : 203-849-7
missing translation for 'rtecsNumber' : SA0320000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1,5-Dibromopentane
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 1,5-Dibromopentane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.