Learn More
(1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) Hexafluorophosphate, 97%, Thermo Scientific Chemicals
(1, 5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate, 97%, C34H38IrP2.F6P, CAS Number-38465-86-0
121.00 € - 279.00 €
Chemical Identifiers
CAS | 38465-86-0 |
---|---|
Molecular Formula | C34H38F6IrP3 |
Molecular Weight (g/mol) | 845.81 |
MDL Number | MFCD00064800 |
InChI Key | LXKHQEQLBSJJCO-JXNOXZOESA-N |
Synonym | 1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate |
PubChem CID | 5702663 |
IUPAC Name | (1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate |
SMILES | [Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10756753
|
Thermo Scientific Acros
377942500 |
250 mg | Glass bottle |
121.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10056172
|
Thermo Scientific Acros
377940010 |
1 g | Glass bottle |
279.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
38465-86-0 | |
845.81 | |
LXKHQEQLBSJJCO-JXNOXZOESA-N | |
5702663 | |
[Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1 |
C34H38F6IrP3 | |
MFCD00064800 | |
1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate | |
(1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate |
Specifications
38465-86-0 | |
100.0 | |
Red | |
96% min | |
MFCD00064800 | |
LXKHQEQLBSJJCO-JXNOXZOESA-N | |
(1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate | |
5702663 | |
97% | |
(1, 5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate |
96.0 | |
224.0°C | |
Authentic | |
C34H38F6IrP3 | |
1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate | |
[Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1 | |
845.81 | |
845.81 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do
GHS Signal Word: Warning
- (1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only