missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methylbenzoyl)pyrrolidine, 97%, Thermo Scientific™

Product Code. 15432468
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59746-40-6
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00996306
InChI Key QXMOFZGGGOFRPH-UHFFFAOYSA-N
Synonym 1-4-methylbenzoyl pyrrolidine, 4-methylphenyl 1-pyrrolidinyl methanone, aronis25719, 4-pyrrolidinecarbonyl toluene, 4-methylphenyl pyrrolidinyl ketone, 1-4-methylphenyl carbonyl pyrrolidine, 4-methylphenyl-pyrrolidin-1-ylmethanone, 4-methylphenyl pyrrolidin-1-yl methanone
PubChem CID 721203
IUPAC Name (4-methylphenyl)-pyrrolidin-1-ylmethanone
SMILES CC1=CC=C(C=C1)C(=O)N2CCCC2

Specifications

Quantity 1 g
Formula Weight 189.25
Percent Purity 97%
Chemical Name or Material 1-(4-Methylbenzoyl)pyrrolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'