missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)ethanol, 95%, Thermo Scientific Chemicals

54.47 € - 174.87 €

Chemical Identifiers

CAS 3319-15-1
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00016857,MFCD09863666,MFCD09863667
InChI Key IUUULXXWNYKJSL-UHFFFAOYNA-N
Synonym 1-4-methoxyphenyl ethanol, 1-4-methoxyphenyl ethan-1-ol, 4-methoxy-alpha-methylbenzyl alcohol, 4-methoxyphenyl methyl carbinol, 1-p-methoxyphenyl ethanol, 4-methoxy-.alpha.-methylbenzyl alcohol, 1-4-methoxyphenyl-ethanol, acmc-20apae, 4-methoxyphenyl ethanol, 1 p-methoxyphenyl ethanol
PubChem CID 101148
ChEBI CHEBI:86566
IUPAC Name 1-(4-methoxyphenyl)ethanol
SMILES COC1=CC=C(C=C1)C(C)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318595
View Documents
Thermo Scientific Alfa Aesar
L12062.14
25 g
54.47 €
25g
Please to purchase this item. Need a web account? Register with us today!
11328595
View Documents
Thermo Scientific Alfa Aesar
L12062.22
100 g
174.87 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-(4-Methoxyphenyl)ethanol is used to study the steady-state and nanosecond, laser-flash photolysis. It is used to produce 4-(1-chloro-ethyl)-anisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methoxyphenyl)ethanol is used to study the steady-state and nanosecond, laser-flash photolysis. It is used to produce 4-(1-chloro-ethyl)-anisole.

Solubility
Not miscible in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

3319-15-1
152.19
IUUULXXWNYKJSL-UHFFFAOYNA-N
101148
1-(4-methoxyphenyl)ethanol
C9H12O2
MFCD00016857,MFCD09863666,MFCD09863667
1-4-methoxyphenyl ethanol, 1-4-methoxyphenyl ethan-1-ol, 4-methoxy-alpha-methylbenzyl alcohol, 4-methoxyphenyl methyl carbinol, 1-p-methoxyphenyl ethanol, 4-methoxy-.alpha.-methylbenzyl alcohol, 1-4-methoxyphenyl-ethanol, acmc-20apae, 4-methoxyphenyl ethanol, 1 p-methoxyphenyl ethanol
CHEBI:86566
COC1=CC=C(C=C1)C(C)O

Specifications

3319-15-1
94°C to 96°C (1 mmHg)
C9H12O2
MFCD00016857,MFCD09863666,MFCD09863667
1-4-methoxyphenyl ethanol, 1-4-methoxyphenyl ethan-1-ol, 4-methoxy-alpha-methylbenzyl alcohol, 4-methoxyphenyl methyl carbinol, 1-p-methoxyphenyl ethanol, 4-methoxy-.alpha.-methylbenzyl alcohol, 1-4-methoxyphenyl-ethanol, acmc-20apae, 4-methoxyphenyl ethanol, 1 p-methoxyphenyl ethanol
IUUULXXWNYKJSL-UHFFFAOYNA-N
1-(4-methoxyphenyl)ethanol
101148
152.19
1-(4-Methoxyphenyl)ethanol
1.079
74°C (165°F)
1.533
2043521
Not miscible in water.
COC1=CC=C(C=C1)C(C)O
152.19
CHEBI:86566
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'einecsNumber' : 222-019-5

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.