missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)piperazine dihydrochloride, 98%, Thermo Scientific™

Product Code. 11343956
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64090-19-3
Molecular Formula C10H15Cl2FN2
Molecular Weight (g/mol) 253.142
MDL Number MFCD00012765
InChI Key DZQVAQAZQDURKX-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl piperazine dihydrochloride, 1-4-fluorophenyl piperazinedihydrochloride, 1-4-fluorophenyl piperazine 2hcl, piperazine, 1-4-fluorophenyl-, dihydrochloride, 1-4-fluorophenyl piperazine;dihydrochloride, para-fluorophenylpiperazine dihydrochloride, zlchem 337, pubchem15283, acmc-209nk7, ksc352o8p
PubChem CID 16211916
IUPAC Name 1-(4-fluorophenyl)piperazine;dihydrochloride
SMILES C1CN(CCN1)C2=CC=C(C=C2)F.Cl.Cl

Specifications

Melting Point ∽234°C (decomposition)
Quantity 500 g
UN Number UN2811
Beilstein 4826840
Formula Weight 253.15
Percent Purity 98%
Chemical Name or Material 1-(4-Fluorophenyl)piperazine dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'