missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™

93.40 € - 286.00 €

Chemical Identifiers

CAS 423768-49-4
Molecular Formula C11H8ClFN2O
Molecular Weight (g/mol) 238.646
MDL Number MFCD03659694
InChI Key INIAJXQDJISNJB-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methylpyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methyl-4-pyrazolecarbonyl chloride, 1-4-fluorophenyl-5-methyl-pyrazole-4-carbonyl chloride
PubChem CID 2779893
IUPAC Name 1-(4-fluorophenyl)-5-methylpyrazole-4-carbonyl chloride
SMILES CC1=C(C=NN1C2=CC=C(C=C2)F)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10364562
View Documents Promotion Details
Thermo Scientific Maybridge
CC00119DA
1 g
93.40 €
1g
Please to purchase this item. Need a web account? Register with us today!
10577264
View Documents Promotion Details
Thermo Scientific Maybridge
CC00119DE
5 g
286.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

423768-49-4
238.646
INIAJXQDJISNJB-UHFFFAOYSA-N
2779893
CC1=C(C=NN1C2=CC=C(C=C2)F)C(=O)Cl
C11H8ClFN2O
MFCD03659694
1-4-fluorophenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methylpyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methyl-4-pyrazolecarbonyl chloride, 1-4-fluorophenyl-5-methyl-pyrazole-4-carbonyl chloride
1-(4-fluorophenyl)-5-methylpyrazole-4-carbonyl chloride

Specifications

423768-49-4
100.0
Amber glass bottle
MFCD03659694
INIAJXQDJISNJB-UHFFFAOYSA-N
1-(4-fluorophenyl)-5-methylpyrazole-4-carbonyl chloride
2779893
97%
96.5
97%
C11H8ClFN2O
1-4-fluorophenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methylpyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methyl-4-pyrazolecarbonyl chloride, 1-4-fluorophenyl-5-methyl-pyrazole-4-carbonyl chloride
CC1=C(C=NN1C2=CC=C(C=C2)F)C(=O)Cl
238.646
238.65
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications