missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluoro-3-methoxyphenyl)ethanol, 95%, Thermo Scientific™

Product Code. 15424807
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870849-56-2
Molecular Formula C9H11FO2
Molecular Weight (g/mol) 170.183
MDL Number MFCD08062372
InChI Key OGBGGCJKDXCBGP-UHFFFAOYSA-N
Synonym 1-4-fluoro-3-methoxyphenyl ethanol, 1-4-fluoro-3-methoxyphenyl ethan-1-ol, benzenemethanol,4-fluoro-3-methoxy-a-methyl
PubChem CID 24721520
IUPAC Name 1-(4-fluoro-3-methoxyphenyl)ethanol
SMILES CC(C1=CC(=C(C=C1)F)OC)O

Specifications

Quantity 1 g
Formula Weight 170.18
Percent Purity 95%
Chemical Name or Material 1-(4-Fluoro-3-methoxyphenyl)ethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'