missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane, CHROMASOLV™ Plus, for HPLC, ≥99.5%, Honeywell Riedel-de Haën™
CHROMASOLV™ Plus, for HPLC,≥99.5%
70.04 € - 3142.69 €
Chemical Identifiers
CAS | 123-91-1 |
---|---|
Molecular Formula | C4H8O2 |
MDL Number | MFCD00006571 |
InChI Key | RYHBNJHYFVUHQT-UHFFFAOYSA-N |
PubChem CID | 31275 |
ChEBI | CHEBI:47032 |
IUPAC Name | 1,4-dioxane |
SMILES | C1COCCO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15634550
|
Honeywell-Chromasolv
34857-100ML |
100 mL | Glass Bottle | ||||||
15674550
|
Honeywell-Chromasolv
34857-500ML |
500 mL | Narrow Mouth Glass Bottle | ||||||
15644550
|
Honeywell-Chromasolv
34857-1L |
1 L | Glass Bottle | ||||||
15654550
|
Honeywell-Chromasolv
34857-2.5L |
2.5 L | Narrow Mouth Glass Bottle |
761.34 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15684550
|
Honeywell-Chromasolv
34857-6X1L |
6 x 1 L | Glass Bottle | ||||||
15664550
|
Honeywell-Chromasolv
34857-4X2.5L |
4 x 2.5 L | Glass Bottle |
3142.69 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Chemical Identifiers
123-91-1 | |
MFCD00006571 | |
31275 | |
1,4-dioxane |
C4H8O2 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
CHEBI:47032 | |
C1COCCO1 |
Specifications
10°C to 12°C (literature) | |
123-91-1 | |
C4H8O2 | |
MFCD00006571 | |
102551 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
1,4-dioxane | |
31275 | |
88.11 | |
HPLC | |
100°C to 102°C | |
1,4-Dioxane, CHROMASOLV™ Plus, for HPLC, ≥99.5% |
max. 0.01 at 295 nm, max. 0.10 at 270 nm, max. 0.20 at 250 nm, max. 0.50 at 235 nm, max. 0.70 at 220 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm | |
100.0000% | |
C4H8O2 | |
UN1165 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
C1COCCO1 | |
88.106 | |
CHEBI:47032 | |
≥99.5% | |
1.034 g/mL at 25 °C(lit.) | |
11°C |
Safety and Handling
missing translation for 'shelfLife' : 720 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Dioxane
missing translation for 'einecsNumber' : 204-661-8
Recommended Storage : Room Temp