Learn More
1,4-Dimethylpiperazine, 98%, Thermo Scientific Chemicals
25.90 € - 58.10 €
Chemical Identifiers
CAS | 106-58-1 |
---|---|
Molecular Formula | C6H14N2 |
Molecular Weight (g/mol) | 114.192 |
MDL Number | MFCD00006156 |
InChI Key | RXYPXQSKLGGKOL-UHFFFAOYSA-N |
Synonym | n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569 |
PubChem CID | 7818 |
IUPAC Name | 1,4-dimethylpiperazine |
SMILES | CN1CCN(CC1)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435877
|
Thermo Scientific Alfa Aesar
B24419.14 |
25 g |
25.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445877
|
Thermo Scientific Alfa Aesar
B24419.22 |
100 g |
58.10 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-58-1 | |
114.192 | |
RXYPXQSKLGGKOL-UHFFFAOYSA-N | |
7818 | |
CN1CCN(CC1)C |
C6H14N2 | |
MFCD00006156 | |
n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569 | |
1,4-dimethylpiperazine |
Specifications
106-58-1 | |
10 | |
21°C (69°F) | |
C6H14N2 | |
UN2733 | |
n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569 | |
CN1CCN(CC1)C | |
114.192 | |
114.19 | |
1,4-Dimethylpiperazine |
-1°C | |
131°C to 132°C | |
Amine-like | |
MFCD00006156 | |
103022 | |
RXYPXQSKLGGKOL-UHFFFAOYSA-N | |
1,4-dimethylpiperazine | |
7818 | |
98% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'rtecsNumber' : TL5945000
missing translation for 'tsca' : TSCA
RUO – Research Use Only