Learn More
1,4-Diethoxybutane, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 13344-00-8 |
---|---|
Molecular Formula | C8H18O2 |
Molecular Weight (g/mol) | 146.23 |
MDL Number | MFCD00048581 |
InChI Key | IIHAWQOFHTYWGM-UHFFFAOYSA-N |
Synonym | butane, 1,4-diethoxy, 1,4-butanediol diethyl ether, butane,4-diethoxy, acmc-1buxo, 1,4-diethoxy butane |
PubChem CID | 232578 |
IUPAC Name | 1,4-diethoxybutane |
SMILES | CCOCCCCOCC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13344-00-8 | |
146.23 | |
IIHAWQOFHTYWGM-UHFFFAOYSA-N | |
232578 | |
CCOCCCCOCC |
C8H18O2 | |
MFCD00048581 | |
butane, 1,4-diethoxy, 1,4-butanediol diethyl ether, butane,4-diethoxy, acmc-1buxo, 1,4-diethoxy butane | |
1,4-diethoxybutane |
Specifications
13344-00-8 | |
48°C (115°F) | |
CH3CH2O(CH2)4OCH2CH3 | |
UN1993 | |
IIHAWQOFHTYWGM-UHFFFAOYSA-N | |
1,4-diethoxybutane | |
232578 | |
98% |
165°C | |
C8H18O2 | |
MFCD00048581 | |
butane, 1,4-diethoxy, 1,4-butanediol diethyl ether, butane,4-diethoxy, acmc-1buxo, 1,4-diethoxy butane | |
CCOCCCCOCC | |
146.23 | |
146.23 | |
1,4-Diethoxybutane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.