Learn More
1,4-Dichlorophthalazine, 98%, Thermo Scientific Chemicals
43.80 € - 142.71 €
Chemical Identifiers
CAS | 4752-10-7 |
---|---|
Molecular Formula | C8H4Cl2N2 |
Molecular Weight (g/mol) | 199.03 |
MDL Number | MFCD00006909 |
InChI Key | ODCNAEMHGMYADO-UHFFFAOYSA-N |
Synonym | phthalazine, 1,4-dichloro, ccris 6799, 1,4-dichloro-phthalazine, unii-0ow60266ko, 1,4-dichlorophthalazine, dichlorophthalazine, 1,4-dichloropthalazine, 1,4-dichorophthalazine, pubchem23238, 1,4-dichlorophtalazine |
PubChem CID | 78490 |
IUPAC Name | 1,4-dichlorophthalazine |
SMILES | C1=CC=C2C(=C1)C(=NN=C2Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15466107
|
Thermo Scientific Alfa Aesar
H33295.03 |
1 g | N/A | N/A | N/A | |||||
15476107
|
Thermo Scientific Alfa Aesar
H33295.06 |
5 g |
142.71 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Chemical Identifiers
4752-10-7 | |
199.03 | |
ODCNAEMHGMYADO-UHFFFAOYSA-N | |
78490 | |
C1=CC=C2C(=C1)C(=NN=C2Cl)Cl |
C8H4Cl2N2 | |
MFCD00006909 | |
phthalazine, 1,4-dichloro, ccris 6799, 1,4-dichloro-phthalazine, unii-0ow60266ko, 1,4-dichlorophthalazine, dichlorophthalazine, 1,4-dichloropthalazine, 1,4-dichorophthalazine, pubchem23238, 1,4-dichlorophtalazine | |
1,4-dichlorophthalazine |
Specifications
4752-10-7 | |
C8H4Cl2N2 | |
Moisture sensitive | |
Sparingly soluble in water.(0.26g/L) (25°C), | |
C1=CC=C2C(=C1)C(=NN=C2Cl)Cl | |
199.03 | |
199.04 | |
1,4-Dichlorophthalazine |
164°C to 166°C | |
MFCD00006909 | |
phthalazine, 1,4-dichloro, ccris 6799, 1,4-dichloro-phthalazine, unii-0ow60266ko, 1,4-dichlorophthalazine, dichlorophthalazine, 1,4-dichloropthalazine, 1,4-dichorophthalazine, pubchem23238, 1,4-dichlorophtalazine | |
ODCNAEMHGMYADO-UHFFFAOYSA-N | |
1,4-dichlorophthalazine | |
78490 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 225-275-6
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.