missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminobutane dihydrochloride, 99+%, Thermo Scientific Chemicals

23.22 € - 197.99 €

Chemical Identifiers

CAS 333-93-7
Molecular Formula C4H12N2·2HCl
Molecular Weight (g/mol) 161.08
MDL Number MFCD00012526
InChI Key XXWCODXIQWIHQN-UHFFFAOYSA-N
Synonym 1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
PubChem CID 9532
IUPAC Name butane-1,4-diamine;dihydrochloride
SMILES C(CCN)CN.Cl.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10430681
View Documents Promotion Details
Thermo Scientific Acros
215110050
5 g Glass bottle
23.22 €
5g
Please to purchase this item. Need a web account? Register with us today!
10175220
View Documents Promotion Details
Thermo Scientific Acros
215110250
25 g Glass bottle
71.96 €
25g
Please to purchase this item. Need a web account? Register with us today!
10229453
View Documents Promotion Details
Thermo Scientific Acros
215111000
100 g Glass bottle
197.99 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

333-93-7
161.08
XXWCODXIQWIHQN-UHFFFAOYSA-N
9532
C(CCN)CN.Cl.Cl
C4H12N2·2HCl
MFCD00012526
1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
butane-1,4-diamine;dihydrochloride

Specifications

333-93-7
100.0
White
99+%
H2N(CH2)4NH2·2HCl
04, 264
1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
XXWCODXIQWIHQN-UHFFFAOYSA-N
butane-1,4-diamine;dihydrochloride
9532
99+%
1, 4-Diaminobutane dihydrochloride
99.0
280.0°C
Authentic
C4H12N2·2HCl
MFCD00012526
15, 8058
Solubility in water: soluble.
C(CCN)CN.Cl.Cl
161.08
161.08
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 206-375-9

Product Identifier
  • 1,4-Diaminobutane dihydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only