missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanedione monoethylene acetal, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.
Solubility
Soluble in chloroform & methanol.
Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents and strong acids. Ensure adequate ventilation during use.
1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.
Solubility
Soluble in chloroform & methanol.
Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents and strong acids. Ensure adequate ventilation during use.
Specifications
Specifications
Melting Point | 72°C to 75°C |
Quantity | 25 g |
Beilstein | 117631 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in chloroform & methanol. |
Formula Weight | 156.18 |
Percent Purity | 97% |
Chemical Name or Material | 1,4-Cyclohexanedione monoethylene acetal |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'