missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanedione, 98%, Thermo Scientific Chemicals
35.00 € - 356.00 €
Chemical Identifiers
CAS | 637-88-7 |
---|---|
Molecular Formula | C6H8O2 |
Molecular Weight (g/mol) | 112.13 |
InChI Key | DCZFGQYXRKMVFG-UHFFFAOYSA-N |
Synonym | 1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar |
PubChem CID | 12511 |
ChEBI | CHEBI:28286 |
IUPAC Name | cyclohexane-1,4-dione |
SMILES | C1CC(=O)CCC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10285480
|
Thermo Scientific Acros
154830100 |
10 g | Glass bottle |
35.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10245530
|
Thermo Scientific Acros
154830500 |
50 g | Glass bottle |
114.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924841
|
Thermo Scientific Acros
154832500 |
250 g | Glass bottle |
356.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
637-88-7 | |
112.13 | |
1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar | |
CHEBI:28286 | |
C1CC(=O)CCC1=O |
C6H8O2 | |
DCZFGQYXRKMVFG-UHFFFAOYSA-N | |
12511 | |
cyclohexane-1,4-dione |
Specifications
637-88-7 | |
Beige or Yellow | |
132°C | |
97.5% min. (GC) | |
C6H8(=O)2 | |
01,178 | |
Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol | |
C1CC(=O)CCC1=O | |
112.13 | |
CHEBI:28286 | |
98% | |
1, 4-Cyclohexanedione |
76.0°C to 80.0°C | |
112°C (26.0 hPa) | |
Authentic | |
C6H8O2 | |
07,556 | |
1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar | |
DCZFGQYXRKMVFG-UHFFFAOYSA-N | |
cyclohexane-1,4-dione | |
12511 | |
112.13 | |
Crystalline Powder or Flakes |
Safety and Handling
missing translation for 'einecsNumber' : 211-306-
RUO – Research Use Only