missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanebis(methylamine), cis + trans, 96%, Thermo Scientific Chemicals

Product Code. 11333037
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2549-93-1
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00039463
InChI Key OXIKYYJDTWKERT-UHFFFAOYSA-N
Synonym 1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanedimethanamine, bamch, cis-1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanebis methylamine, cyclohexane-1,4-diyldimethanamine, trans-1,4-bis aminomethyl cyclohexane, 4-aminomethyl cyclohexyl methanamine, trans-1,4-cyclohexanedimethanamine, cyclohex-1,4-ylenebis methylamine
PubChem CID 17354
IUPAC Name [4-(aminomethyl)cyclohexyl]methanamine
SMILES C1CC(CCC1CN)CN

Specifications

Density 0.95
Boiling Point 239°C to 244°C
Flash Point 106°C (222°F)
Refractive Index 1.49
Quantity 5 g
UN Number UN2922
Beilstein 2071383
Sensitivity Air Sensitive
Formula Weight 142.25
Percent Purity 96%
Chemical Name or Material 1,4-Cyclohexanebis(methylamine), Cis + Trans
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.