missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol, 99%, Thermo Scientific Chemicals

24.42 € - 81.10 €

Chemical Identifiers

CAS 110-63-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002968
InChI Key WERYXYBDKMZEQL-UHFFFAOYSA-N
Synonym 1,4-butanediol, 1,4-butylene glycol, tetramethylene glycol, 1,4-dihydroxybutane, 1,4-tetramethylene glycol, tetramethylene 1,4-diol, sucol b, diol 14b, agrisynth b1d, unii-7xoo2le6g3
PubChem CID 8064
ChEBI CHEBI:41189
IUPAC Name butane-1,4-diol
SMILES OCCCCO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11382176
View Documents
Thermo Scientific Alfa Aesar
L03491.30
250 g
24.42 €
250g
Please to purchase this item. Need a web account? Register with us today!
11456720
View Documents
Thermo Scientific Alfa Aesar
L03491.0B
1000 g
40.60 €
1000g
Please to purchase this item. Need a web account? Register with us today!
11466720
View Documents
Thermo Scientific Alfa Aesar
L03491.0E
2500 g
81.10 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,4-Butanediol is commonly used as a solvent in the chemical industry to manufacture gamma-butyrolactone and elastic fibers like spandex. It is used as a cross-linking agent for thermoplastic urethanes, polyester plasticizers, paints and coatings. It undergoes dehydration in the presence of phosphoric acid yielded teterahydrofuran, which is an important solvent used for various applications. It acts an intermediate and is used to manufacture polytetramethylene ether glycol (PTMEG), polybutylene terephthalate (PBT) and polyurethane (PU). It finds application as an industrial cleaner and a glue remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Butanediol is commonly used as a solvent in the chemical industry to manufacture gamma-butyrolactone and elastic fibers like spandex. It is used as a cross-linking agent for thermoplastic urethanes, polyester plasticizers, paints and coatings. It undergoes dehydration in the presence of phosphoric acid yielded teterahydrofuran, which is an important solvent used for various applications. It acts an intermediate and is used to manufacture polytetramethylene ether glycol (PTMEG), polybutylene terephthalate (PBT) and polyurethane (PU). It finds application as an industrial cleaner and a glue remover.

Solubility
Miscible with water, ethanol, dimethyl sulfoxide and glycol ethers. Slightly miscible with diethyl ether. Immiscible with aliphatic and aromatic hydrocarbons.

Notes
Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and reducing agents.
Specifications

Chemical Identifiers

110-63-4
90.12
WERYXYBDKMZEQL-UHFFFAOYSA-N
8064
butane-1,4-diol
C4H10O2
MFCD00002968
1,4-butanediol, 1,4-butylene glycol, tetramethylene glycol, 1,4-dihydroxybutane, 1,4-tetramethylene glycol, tetramethylene 1,4-diol, sucol b, diol 14b, agrisynth b1d, unii-7xoo2le6g3
CHEBI:41189
OCCCCO

Specifications

110-63-4
1.017
135°C (275°F)
1.446
1633445
1,4-butanediol, 1,4-butylene glycol, tetramethylene glycol, 1,4-dihydroxybutane, 1,4-tetramethylene glycol, tetramethylene 1,4-diol, sucol b, diol 14b, agrisynth b1d, unii-7xoo2le6g3
WERYXYBDKMZEQL-UHFFFAOYSA-N
butane-1,4-diol
8064
90.12
1,4-Butanediol
19°C to 20°C
229°C to 230°C
C4H10O2
MFCD00002968
Hygroscopic
Miscible with water,ethanol,dimethyl sulfoxide and glycol ethers. Slightly miscible with diethyl ether. Immiscible with aliphatic and aromatic hydrocarbons.
OCCCCO
90.12
CHEBI:41189
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302-H336

missing translation for 'einecsNumber' : 203-786-5

missing translation for 'rtecsNumber' : EK0525000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • 1,4-Butanediol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Supplemental information
  • MIXTURE LIST-Contains : 1,4-Butanediol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only