missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trimethylsilyl)tetrafluorobenzene, 98%, Thermo Scientific Chemicals

Product Code. 15453337
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 53°C to 55°C
CAS 16956-91-5
Quantity 1 g
Molecular Formula C12H18F4Si2
MDL Number MFCD00093109
Synonym 1,4-bis trimethylsilyl tetrafluorobenzene, trimethyl 2,3,5,6-tetrafluoro-4-trimethylsilyl phenyl silane, acmc-20ap2x, 1,4-bis-trimethylsilyl tetrafluorobenzene, perfluoro-1,4-phenylene bis trimethylsilane, tetrafluoro-1,4-phenylenebis trimethylsilane, 1,2,4,5-tetrafluoro-3,6-bis trimethylsilyl-benzene, benzene,1,2,4,5-tetrafluoro-3,6-bis trimethylsilyl, trimethyl-2,3,5,6-tetrafluoro-4-trimethylsilylphenyl silane
InChI Key RIGHSQASRZUCPK-UHFFFAOYSA-N
SMILES C[Si](C)(C)C1=C(C(=C(C(=C1F)F)[Si](C)(C)C)F)F
IUPAC Name trimethyl-(2,3,5,6-tetrafluoro-4-trimethylsilylphenyl)silane
Molecular Weight (g/mol) 294.44
PubChem CID 2769673
Formula Weight 294.44
Percent Purity 99%
Chemical Name or Material 1,4-Bis(trimethylsilyl)tetrafluorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'