missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzoquinone, 99%, Thermo Scientific Chemicals

Product Code. 10073713
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

1, 4-Benzoquinone, 99%, C6H4O2, CAS Number-106-51-4, 1,4-benzoquine, 1,4-benzoquinone, 2,5-cyclohexadiene-1,4-dione, benzoquinone, chinone, cyclohexadienedione, para-quinone, p-benzoquinone, p-quinone, quinone, 1kg, 98.5% min. (Iodometry), 07,609, 100.0, CHEBI:16509, Yellow to Green, 1.3100g/mL

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-51-4
Molecular Formula C6H4O2
Molecular Weight (g/mol) 108.1
MDL Number MFCD00001591
InChI Key AZQWKYJCGOJGHM-UHFFFAOYSA-N
Synonym p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine
PubChem CID 4650
ChEBI CHEBI:16509
IUPAC Name cyclohexa-2,5-diene-1,4-dione
SMILES C1=CC(=O)C=CC1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 112°C to 116°C
Color Yellow to Green
Density 1.3100g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Plastic drum
Linear Formula C6H4(=O)2
Quantity 5 kg
Beilstein 07,609
Fieser 01,49; 07,55
Merck Index 15,8191
Specific Gravity 1.31
Solubility Information Solubility in water: 10g/L water (25°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
Formula Weight 108.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 4-Benzoquinone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.