Learn More
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
Brand:  Trajan Scientific and Medical™ 0624313
 View more versions of this product
Product Code. 15238773
  • 46.45 € / Pack of 2
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Specifications
Tubing
2/Pk.
0.1 mm
150 mm