missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(Trifluoromethoxy)phenyl]ethanol, 97%, Thermo Scientific™

Product Code. 15405117
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 347194-02-9
Molecular Formula C9H9F3O2
Molecular Weight (g/mol) 206.164
MDL Number MFCD06797343
InChI Key BWANKNAGTRWECQ-UHFFFAOYSA-N
Synonym 1-3-trifluoromethoxy phenyl ethanol, 1-3-trifluoromethoxy phenyl ethan-1-ol, 1-3-trifluoromethoxyphenyl ethanol
PubChem CID 44891072
IUPAC Name 1-[3-(trifluoromethoxy)phenyl]ethanol
SMILES CC(C1=CC(=CC=C1)OC(F)(F)F)O

Specifications

Quantity 25 g
Formula Weight 206.16
Percent Purity 97%
Chemical Name or Material 1-[3-(Trifluoromethoxy)phenyl]ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.