Learn More
1,3-Indanedione, 97%, Thermo Scientific Chemicals
33.97 € - 319.59 €
Chemical Identifiers
CAS | 606-23-5 |
---|---|
Molecular Formula | C9H6O2 |
Molecular Weight (g/mol) | 146.145 |
MDL Number | MFCD00003779 |
InChI Key | UHKAJLSKXBADFT-UHFFFAOYSA-N |
Synonym | 1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g |
PubChem CID | 11815 |
ChEBI | CHEBI:78877 |
IUPAC Name | indene-1,3-dione |
SMILES | C1C(=O)C2=CC=CC=C2C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11464873
|
Thermo Scientific Alfa Aesar
A12206.06 |
5 g |
33.97 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474873
|
Thermo Scientific Alfa Aesar
A12206.14 |
25 g |
109.55 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484873
|
Thermo Scientific Alfa Aesar
A12206.22 |
100 g |
319.59 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.
Solubility
Soluble in ethanol, ether, benzene, slightly soluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Chemical Identifiers
606-23-5 | |
146.145 | |
UHKAJLSKXBADFT-UHFFFAOYSA-N | |
11815 | |
indene-1,3-dione |
C9H6O2 | |
MFCD00003779 | |
1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g | |
CHEBI:78877 | |
C1C(=O)C2=CC=CC=C2C1=O |
Specifications
606-23-5 | |
1.37 | |
MFCD00003779 | |
1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g | |
UHKAJLSKXBADFT-UHFFFAOYSA-N | |
indene-1,3-dione | |
11815 | |
146.15 | |
1,3-Indanedione |
128°C to 132°C | |
C9H6O2 | |
1210061 | |
Soluble in ethanol,ether,benzene,slightly soluble in water. | |
C1C(=O)C2=CC=CC=C2C1=O | |
146.145 | |
CHEBI:78877 | |
97% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319
missing translation for 'einecsNumber' : 210-109-7
missing translation for 'rtecsNumber' : NK5070000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1,3-Indanedione
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : 1H-Indene-1,3(2H)-dione
RUO – Research Use Only