missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dioxolane, ReagentPlus™, 99%, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
Glass Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 1.06g/mL at 25°C |
Boiling Point | 76°C |
Flash Point | −3°C (26.6°F) |
Ignition Point | 525°F |
Packaging | Glass Bottle |
Refractive Index | n20/D 1.401(lit.) |
Quantity | 1 L |
UN Number | UN1166 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'