missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, ReagentPlus™, 99%, Honeywell™

Product Code. 15608330
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, contains ∼75 ppm BHT as inhibitor, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-06-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 12586
ChEBI CHEBI:87597
IUPAC Name 1,3-dioxolane
SMILES C1COCO1

Specifications

Melting Point -95°C
Color Colorless
Density 1.06g/mL at 25°C
Boiling Point 76°C
Flash Point −3°C (26.6°F)
Ignition Point 525°F
Packaging Aluminum Bottle
Refractive Index n20/D 1.401(lit.)
Quantity 4 L
UN Number UN1166
Beilstein 102453
Formula Weight 74.08g/mol
Vapor Pressure 70mmHg (20°C)
Percent Purity 99%
Grade ReagentPlus™
Chemical Name or Material 1,3-Dioxolane, Contains approx.75 ppm BHT as inhibitor
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'