missing translation for 'onlineSavingsMsg'
Learn More

1-(3,5-Difluoro-2-hydroxyphenyl)ethan-1-one, Thermo Scientific™

29.00 € - 224.00 €

Chemical Identifiers

CAS 140675-42-9
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00042476
InChI Key MCDJUVXLLXTCFP-UHFFFAOYSA-N
Synonym 1-3,5-difluoro-2-hydroxyphenyl ethanone, 1-3,5-difluoro-2-hydroxyphenyl ethan-1-one, 3',5'-difluoro-2'-hydroxyacetophenone, 3,5-difluoro-2-hydroxyacetophenone, 2-acetyl-4,6-difluorophenol, ethanone,1-3,5-difluoro-2-hydroxyphenyl, 1-acetyl-3,5-difluoro-2-hydroxybenzene, acmc-1byqa, 1-3,5-difluoro-2-hydroxyphenyl-ethanone, 1-3,5-difluoro-2-hydroxy-phenyl-ethanone
PubChem CID 2736976
IUPAC Name 1-(3,5-difluoro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1O)F)F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10262772
View Documents
Thermo Scientific Maybridge
BTB04131DA
1 g Amber Glass Bottle
29.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10272772
View Documents
Thermo Scientific Maybridge
BTB04131EA
10 g Amber Glass Bottle
224.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

140675-42-9
172.131
MCDJUVXLLXTCFP-UHFFFAOYSA-N
2736976
CC(=O)C1=CC(=CC(=C1O)F)F
C8H6F2O2
MFCD00042476
1-3,5-difluoro-2-hydroxyphenyl ethanone, 1-3,5-difluoro-2-hydroxyphenyl ethan-1-one, 3',5'-difluoro-2'-hydroxyacetophenone, 3,5-difluoro-2-hydroxyacetophenone, 2-acetyl-4,6-difluorophenol, ethanone,1-3,5-difluoro-2-hydroxyphenyl, 1-acetyl-3,5-difluoro-2-hydroxybenzene, acmc-1byqa, 1-3,5-difluoro-2-hydroxyphenyl-ethanone, 1-3,5-difluoro-2-hydroxy-phenyl-ethanone
1-(3,5-difluoro-2-hydroxyphenyl)ethanone

Specifications

140675-42-9
100.0
C8H6F2O2
1-3,5-difluoro-2-hydroxyphenyl ethanone, 1-3,5-difluoro-2-hydroxyphenyl ethan-1-one, 3',5'-difluoro-2'-hydroxyacetophenone, 3,5-difluoro-2-hydroxyacetophenone, 2-acetyl-4,6-difluorophenol, ethanone,1-3,5-difluoro-2-hydroxyphenyl, 1-acetyl-3,5-difluoro-2-hydroxybenzene, acmc-1byqa, 1-3,5-difluoro-2-hydroxyphenyl-ethanone, 1-3,5-difluoro-2-hydroxy-phenyl-ethanone
CC(=O)C1=CC(=CC(=C1O)F)F
172.131
172.13
1-(3, 5-Difluoro-2-hydroxyphenyl)ethan-1-one
96.5
97%
MFCD00042476
MCDJUVXLLXTCFP-UHFFFAOYSA-N
1-(3,5-difluoro-2-hydroxyphenyl)ethanone
2736976
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.