missing translation for 'onlineSavingsMsg'
Learn More

1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline), 99%, Thermo Scientific Chemicals

Product Code. 10617231
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 23001-29-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 149°C to 150°C
Color Brown to Purple
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C19H19NO2
MDL Number MFCD00051350
Quantity 5 g
Synonym 1',3',3'-trimethyl-6-hydroxyspiro 2h-1-benzopyran-2,2'-indoline, 1',3',3'-trimethylspiro chromene-2,2'-indolin-6-ol, spiro 2h-1-benzopyran-2,2'-indolin-6-ol, 1',3',3'-trimethyl, spiro 2h-1-benzopyran-2,2'-2h indol-6-ol, 1',3'-dihydro-1',3',3'-trimethyl, 1',3',3'-trimethylspiro chromene-2,2'-indole-6-ol, 1',3',3'-trimethylspiro chromene-2,2'-indol-6-ol, bio2g2, 1',3',3'-trimethylspiro chromene-2,2'-indoline-6-ol
InChI Key UHRBDHPBCHWWAG-UHFFFAOYNA-N
SMILES CN1C2=CC=CC=C2C(C)(C)C11OC2=CC=C(O)C=C2C=C1
IUPAC Name 1',3',3'-trimethylspiro[chromene-2,2'-indole]-6-ol
Molecular Weight (g/mol) 293.37
PubChem CID 90866
Formula Weight 293.36
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline), 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.