missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Trimethylsilylethoxy)methyl-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

169.85 € - 390.00 €

Chemical Identifiers

CAS 903550-12-9
Molecular Formula C15H29BN2O3Si
Molecular Weight (g/mol) 324.303
MDL Number MFCD09037502
InChI Key FVCZMGOMGHMAIX-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl-1h-pyrazole, 1-2-trimethylsilylethoxy methylpyrazole-5-boronic acid, pinacol ester, 1-2-trimethylsilanylethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxy methyl-1h-pyrazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl pyrazole, 1-2-trimethylsilyl ethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxyl methyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilyl ethoxymethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1h-pyrazole, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1-2-trimethylsilanyl-ethoxymethyl-1h-pyrazole
PubChem CID 44755201
IUPAC Name trimethyl-[2-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methoxy]ethyl]silane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2COCC[Si](C)(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15495115
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H27043.MD
250 mg
169.85 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11496070
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H27043.03
1 g
390.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

903550-12-9
324.303
FVCZMGOMGHMAIX-UHFFFAOYSA-N
44755201
B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2COCC[Si](C)(C)C
C15H29BN2O3Si
MFCD09037502
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl-1h-pyrazole, 1-2-trimethylsilylethoxy methylpyrazole-5-boronic acid, pinacol ester, 1-2-trimethylsilanylethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxy methyl-1h-pyrazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl pyrazole, 1-2-trimethylsilyl ethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxyl methyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilyl ethoxymethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1h-pyrazole, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1-2-trimethylsilanyl-ethoxymethyl-1h-pyrazole
trimethyl-[2-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methoxy]ethyl]silane

Specifications

903550-12-9
MFCD09037502
FVCZMGOMGHMAIX-UHFFFAOYSA-N
trimethyl-[2-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methoxy]ethyl]silane
44755201
95%
C15H29BN2O3Si
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl-1h-pyrazole, 1-2-trimethylsilylethoxy methylpyrazole-5-boronic acid, pinacol ester, 1-2-trimethylsilanylethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxy methyl-1h-pyrazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl pyrazole, 1-2-trimethylsilyl ethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxyl methyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilyl ethoxymethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1h-pyrazole, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1-2-trimethylsilanyl-ethoxymethyl-1h-pyrazole
B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2COCC[Si](C)(C)C
324.303
324.31
1-(2-Trimethylsilylethoxy)methyl-1H-pyrazole-5-boronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only